Learn More
3M™ Egg White Protein Rapid Kit L25EGG, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. s26911 Shop All AquaPhoenix Scientific Products
Click to view available options
:
Egg White Protein Rapid Kit L25EGG, Sold by AquaPhoenix Scientific
For screening for the presence of egg white proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products
  • Results visible at 11 ± 1 min. after application of sample
  • Rapid, qualitative immunochromatographic lateral flow test cartridges designed for detection of allergenic food proteins
  • Designed to detect both processed and unprocessed allergen proteins
  • Test can accommodate environmental swab, CIP rinse water and food samples
Specifications
Format Kit
Product Type Egg White Protein Test Kit
Test Type Allergen
For Use With (Application) Screening egg white proteins in CIP final rinse water, environmental swab samples, food ingredients and processed food products
Quantity 25/Cs.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.