Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN1/PHD2 Antibody (366G/76/3), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP130328IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130328IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130328IR Supplier Novus Biologicals Supplier No. NBP130328IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EGLN1/PHD2 Monoclonal specifically detects EGLN1/PHD2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EGLN1/PHD2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 366G/76/3
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179
Gene Symbols EGLN1
Host Species Mouse
Immunogen A peptide within residues 1-50 of human PHD2/HIF Prolyl Hydroxylase 2. [Swiss-Prot# Q9GZT9]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Chromatin Research, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 54583
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.