Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN1/PHD2 Antibody (366G/76/3), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB392153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392153 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392153 Supplier Novus Biologicals Supplier No. NBP308507JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EGLN1/PHD2 Monoclonal antibody specifically detects EGLN1/PHD2 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen EGLN1/PHD2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 366G/76/3
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias C1orf12, EC 1.14.11.29, EC:1.14.11.291, ECYT3, egl nine homolog 1, egl nine-like protein 1, EGLN1, HALAH, HIF prolyl hydroxylase 2, HIFPH2, HIF-PH2, HIF-prolyl hydroxylase 2, HPH2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2, PNAS-118, PNAS-137, Prolyl hydroxylase domain-containing protein 2, SM20, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6
Host Species Mouse
Immunogen Residues 1-24 of EGLN1/PHD2 (Uniprot: Q9GZT9)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Hypoxia, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 54583
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.