Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN1/PHD2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB100138F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100138F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100138F Supplier Novus Biologicals Supplier No. NB100138F

Rabbit Polyclonal Antibody

EGLN1/PHD2 Polyclonal specifically detects EGLN1/PHD2 in Human, Rat, Mouse (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen EGLN1/PHD2
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Concentration 1mg/mL
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9GZT9
Gene Alias C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179
Gene Symbols EGLN1
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of human PHD2/HIF Prolyl Hydroxylase 2 (between amino acids 350-426) (GeneID 54583).
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 54583
Test Specificity Human. Does not appear to work in mouse. Rat reactivity reported in scientific literature (PMID: 17003483).
Target Species Human, Rat, Mouse (Negative)
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.