Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN3/PHD3 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB100303X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100303X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100303X Supplier Novus Biologicals Supplier No. NB100303AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EGLN3/PHD3 Polyclonal specifically detects EGLN3/PHD3 in Human, Mouse, Primate, Rat samples. It is validated for ChIP Assay, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen EGLN3/PHD3
Applications ChIP Assay, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9H6Z9
Gene Alias EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3
Gene Symbols EGLN3
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of humanPHD3/HIF Prolyl Hydroxylase 3. [LocusLink ID 112399]
Molecular Weight of Antigen 27 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chromatin Research, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 112399
Test Specificity Human and mouse. Primate reactivity reported in scientific literature (PMID: 23732909). mmunogen sequence has 92% homology to rat.
Target Species Human, Mouse, Primate, Primate, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.