Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN3/PHD3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100139 0.1 mL
NB100139SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100139 Supplier Novus Biologicals Supplier No. NB100139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 46 publications

EGLN3/PHD3 Polyclonal specifically detects EGLN3/PHD3 in Human, Mouse, Rat samples. It is validated for ChIP Assay, Electron Microscopy, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Mass Spectrometry, Western Blot.

Specifications

Antigen EGLN3/PHD3
Applications ChIP Assay, Electron Microscopy, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Mass Spectrometry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Chromatin Immunoprecipitation reported in scientific literature (PMID 24367580), Immunohistochemistry 1:250-1:1000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:250-1:1000, Electron Microscopy reported in scientific literature (PMID 17003483), Mass Spectrometry, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9H6Z9
Gene Alias EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3
Gene Symbols EGLN3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues between 50-100 of human PHD3/HIF Prolyl Hydroxylase 3 using the numbering given in entry NP_071356.1 (GeneID 112399).
Molecular Weight of Antigen 28 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 112399
Test Specificity Human and rat. Mouse reactivity reported in scientific literature (PMID: 24037093).
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.