Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGLN3/PHD3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 46 publications
$243.50 - $499.50
Specifications
Antigen | EGLN3/PHD3 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000-1:2000, Chromatin Immunoprecipitation reported in scientific literature (PMID 24367580), Immunohistochemistry 1:250-1:1000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:250-1:1000, Electron Microscopy reported in scientific literature (PMID 17003483), Mass Spectrometry, KnockDown Validated |
Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100139SS
![]() |
Novus Biologicals
NB100139SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB100139
![]() |
Novus Biologicals
NB100139 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
EGLN3/PHD3 Polyclonal specifically detects EGLN3/PHD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy, Mass Spectrometry, Knockdown Validated.Specifications
EGLN3/PHD3 | |
Western Blot 1:1000-1:2000, Chromatin Immunoprecipitation reported in scientific literature (PMID 24367580), Immunohistochemistry 1:250-1:1000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:250-1:1000, Electron Microscopy reported in scientific literature (PMID 17003483), Mass Spectrometry, KnockDown Validated | |
Polyclonal | |
Rabbit | |
Cancer, Hypoxia | |
PBS with 0.05% Sodium Azide | |
EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 | |
EGLN3 | |
IgG | |
Affinity Purified | |
Human and rat. Mouse reactivity reported in scientific literature (PMID: 24037093). |
1 mg/mL | |
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Q9H6Z9 | |
112399 | |
Synthetic peptide corresponding to residues between 50-100 of human PHD3/HIF Prolyl Hydroxylase 3 using the numbering given in entry NP_071356.1 (GeneID 112399). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
28 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title