Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN3/PHD3 Antibody (EG188e/d5), HRP, Novus Biologicals™
SDP

Catalog No. NBP130440H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130440H 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130440H Supplier Novus Biologicals Supplier No. NBP130440H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EGLN3/PHD3 Monoclonal specifically detects EGLN3/PHD3 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EGLN3/PHD3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EG188e/d5
Conjugate HRP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. Q9H6Z9
Gene Alias EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3
Gene Symbols EGLN3
Host Species Mouse
Immunogen Partial recombinant human PHD3/HIF Prolyl Hydroxylase 3 (UniProt# Q9H6Z9)
Molecular Weight of Antigen 27 kDa
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chromatin Research, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 112399
Test Specificity Human, monkey and rat. Immunogen sequence has 98% identity to bovine and mouse.
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.