Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGR1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP178775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178775 Supplier Novus Biologicals Supplier No. NBP178775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EGR1 Polyclonal specifically detects EGR1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP).

Specifications

Antigen EGR1
Applications ChIP Assay, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunoprecipitation 2-10 ug/mg lysate, Chromatin Immunoprecipitation (ChIP)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. P08046
Gene Alias AT225Transcription factor ETR103, early growth response 1, early growth response protein 1, EGR-1, G0S30, KROX24, KROX-24, Nerve growth factor-induced protein A, NGFI-ATranscription factor Zif268, TIS8, ZIF-268, Zinc finger protein Krox-24, ZNF225Zinc finger protein 225
Gene Symbols EGR1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of mouse Early Growth Response Protein 1 using the numbering given in entry NP_031939.1 (GeneID 13653).
Molecular Weight of Antigen 56 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1958
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Rhesus Monkey, Gorilla, Tasmanian devil, Africanelephant, Gray short-tailed opossum, Small-eared galago and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.