Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGR2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1109723A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1109723A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1109723A32 Supplier Novus Biologicals Supplier No. NB11059723AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EGR2 Polyclonal specifically detects EGR2 in Human, Mouse, Porcine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen EGR2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AT591, CMT1D, CMT4E, early growth response 2, early growth response 2 (Krox-20 homolog, Drosophila), early growth response protein 2, EGR-2, FLJ14547, Krox-20 homolog, Drosophila, KROX-20, Drosophila, homolog (early growth response-2), KROX20DKFZp686J1957, Zinc finger protein Krox-20
Gene Symbols EGR2
Host Species Rabbit
Immunogen A synthetic peptide made to a portion of human EGR2 (within residues 200-300). [Swiss-ProtNo. P11161]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1959
Target Species Human, Mouse, Pig
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.