Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHHADH Antibody (OTI2C4), PE, Novus Biologicals™
SDP

Catalog No. NB289466 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289466 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289466 Supplier Novus Biologicals Supplier No. NBP270625PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EHHADH Monoclonal antibody specifically detects EHHADH in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen EHHADH
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C4
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase, enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase, LBFP, LBP, L-PBE, PBFEMGC120586, peroxisomal, peroxisomal bifunctional enzyme3,2-trans-enoyl-CoA isomerase, peroxisomal enoyl-CoA hydratase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 496-723 of human EHHADH (NP_001957) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1962
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.