Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EHMT2 Mouse anti-Human, Mouse, Clone: 1A12E3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | EHMT2 |
|---|---|
| Clone | 1A12E3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7106
![]()
|
Proteintech Group Inc
666891IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.Specifications
| EHMT2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BAT8, C6orf30, DKFZp686H08213, EHMT2, FLJ35547, G9A, H3 K9 HMTase 3, HLA B associated transcript 8, KMT1C, Lysine N methyltransferase 1C, NG36, NG36/G9a, Protein G9a | |
| EHMT2 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1A12E3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q96KQ7, Q9Z148 | |
| 10919, 110147 | |
| EHMT2 Fusion Protein Ag27477 | |
| Primary | |
| -20°C | |
| EHMT2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title