Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eicosane, 99%

Catalog No. AC204375000
Click to view available options
Quantity:
2 g
100 g
500 g
Packaging:
Glass bottle

CAS: 112-95-8 | C20H42 | 282.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-95-8
Molecular Formula C20H42
Molecular Weight (g/mol) 282.56
MDL Number MFCD00009344
InChI Key CBFCDTFDPHXCNY-UHFFFAOYSA-N
Synonym eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane
PubChem CID 8222
ChEBI CHEBI:43619
IUPAC Name icosane
SMILES CCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 36.0°C to 38.0°C
Color White
Density 0.7886g/mL
Boiling Point 220.0°C (30.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)18CH3
Quantity 500 g
Beilstein 01, 174
Specific Gravity 0.7886
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether
Formula Weight 282.54
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Eicosane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
EINECSNumber 204-018-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.