Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EID1 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB044675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044675 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044675 Supplier Novus Biologicals Supplier No. NBP297540FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EID1 Polyclonal specifically detects EID1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EID1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 21 kDa pRb-associated protein, C15orf3PTD014, CREBBP/EP300 inhibitor 1, CREBBP/EP300 inhibitory protein 1MGC138884, CRI1EP300-interacting inhibitor of differentiation 1, E1A-like inhibitor of differentiation 1, EID-1MGC138883, EP300 interacting inhibitor of differentiation 1, IRO45620, NB4 apoptosis related protein, Rb- and p300-binding protein EID-1, RBP21, retinoblastoma protein-associated protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human EID1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 23741
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.