Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF1AY Antibody (OTI2G2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N271417A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271417A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271417A405 Supplier Novus Biologicals Supplier No. NBP271417AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EIF1A Monoclonal specifically detects EIF1A in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen EIF1A
Applications Western Blot
Classification Monoclonal
Clone OTI2G2
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias eIF-1A, EIF1AP1, eIF-4C, EIF4C, eukaryotic translation initiation factor 1A, X chromosome, eukaryotic translation initiation factor 1A, X-chromosomal, eukaryotic translation initiation factor 1A, Y chromosome, eukaryotic translation initiation factor 1A, Y-chromosomal, Eukaryotic translation initiation factor 4C, FLJ45762, Putative eukaryotic translation initiation factor 1A
Gene Symbols EIF1AX
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-144 of human EIF1AY (NP_004672) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 1964
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.