Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF1AY Antibody (OTI2G2), PerCP, Novus Biologicals™
SDP

Catalog No. NB288870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288870 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288870 Supplier Novus Biologicals Supplier No. NBP271417PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EIF1AY Monoclonal antibody specifically detects EIF1AY in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen EIF1AY
Applications Western Blot
Classification Monoclonal
Clone OTI2G2
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias eIF-1A Y isoform, eIF-4C, eukaryotic translation initiation factor 1A, Y chromosome, eukaryotic translation initiation factor 1A, Y-chromosomal, eukaryotic translation initiation factor 1A, Y-linked, Eukaryotic translation initiation factor 4C
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-144 of human EIF1AY(NP_004672) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 9086
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.