Learn More
Description
Specifications
Specifications
| Antigen | eIF2 alpha/EIF2S1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 50000000000 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:100-1:2000 |
| Formulation | PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | NP_004085 |
| Gene Alias | EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
