Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF2 alpha/EIF2S1 Antibody (5E10), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB310068 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310068 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310068 Supplier Novus Biologicals Supplier No. NBP197936MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eIF2 alpha/EIF2S1 Monoclonal antibody specifically detects eIF2 alpha/EIF2S1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen eIF2 alpha/EIF2S1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 5E10
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa
Host Species Mouse
Immunogen Recombinant human eIF2 alpha/EIF2S1 (1-315aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, Lipid and Metabolism, Phospho Specific, Translation Control, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 1965
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.