Learn More
eIF2 alpha/EIF2S1 Antibody (OTI3H4), Janelia Fluor™ 549, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | eIF2 alpha/EIF2S1 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI3H4 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa |
| Gene Symbols | EIF2S1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.