Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF2 alpha/EIF2S1, p Ser51 Antibody (1C6), Novus Biologicals™
SDP

Catalog No. NB298123 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298123 100 μL
NB298122 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298123 Supplier Novus Biologicals Supplier No. NBP326427100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

eIF2 alpha/EIF2S1 Monoclonal antibody specifically detects eIF2 alpha/EIF2S1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen eIF2 alpha/EIF2S1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1C6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa
Host Species Rabbit
Immunogen A synthesized peptide derived from Human eIF2 alpha/EIF2S1 [p Ser51] [UniProt P05198]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, Lipid and Metabolism, Phospho Specific, Translation Control, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 1965
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.