Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF2B3 Antibody (OTI1A4), PE, Novus Biologicals™
SDP

Catalog No. NB288888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288888 Supplier Novus Biologicals Supplier No. NBP271397PE

Mouse Monoclonal Antibody

eIF2B3 Monoclonal antibody specifically detects eIF2B3 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen eIF2B3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI1A4
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EIF-2B, eIF-2B GDP-GTP exchange factor subunit gamma, EIF2Bgamma, eukaryotic translation initiation factor 2B, subunit 3 (gamma, 58kD), eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa, translation initiation factor eIF-2B subunit gamma
Host Species Mouse
Immunogen Full length human recombinant protein of human EIF2B3(NP_065098) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8891
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.