Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF2d Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | eIF2d |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119100
|
Invitrogen™
PA5119100 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122016
|
Invitrogen™
PA5122016 |
20 μL |
Each for $237.00
|
|
|||||
Description
eIF2d Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a translation initiation factor involved in the recruitment and delivery of aminoacyl-tRNAs to the P-site of the eukaryotic ribosome in a GTP-independent manner. This gene was previously referred to as ligatin, but is now known to localize to the cytoplasm and localize and function with translation factors. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
eIF2d | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
EIF2D | |
D1Ertd5e; Eif2d; Eukaryotic translation initiation factor 2D; HCA56; Hepatocellular carcinoma-associated antigen 56; LGTN; Ligatin | |
EIF2D | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q61211 | |
16865 | |
Recombinant protein Eukaryotic translation initiation factor 2D. The antigen corresponds to amino acid range 360-570 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title