Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2G Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093641 20 μL
NB093640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093641 Supplier Novus Biologicals Supplier No. NBP30351320UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EIF2G Polyclonal specifically detects EIF2G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EIF2G
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EIF2, eIF-2gA, EIF2gamma, eIF-2-gamma X, EIF2Geukaryotic translation initiation factor 2, subunit 3 (gamma, 52kD), eIF-2gX, eukaryotic translation initiation factor 2 subunit 3, Eukaryotic translation initiation factor 2 subunit gamma X, eukaryotic translation initiation factor 2, subunit 3 gamma, 52kDa, eukaryotic translation initiation factor 2G
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 173-472 of human EIF2G (NP_001406.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1968
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.