Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3B Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB330253 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB330253 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB330253 Supplier Novus Biologicals Supplier No. NBP297742F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EIF3B Polyclonal antibody specifically detects EIF3B in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen EIF3B
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias eIF3 p110, eIF3 p116, eIF3b, eIF-3-eta, EIF3-P110, EIF3-P116, EIF3S9EIF3-ETA, Eukaryotic translation initiation factor 3 subunit 9, eukaryotic translation initiation factor 3 subunit B, eukaryotic translation initiation factor 3, subunit 9 (eta, 116kD), eukaryotic translation initiation factor 3, subunit 9 eta, 116kDa, eukaryotic translation initiation factor 3, subunit B, MGC104664, MGC131875, Prt1 homolog, PRT1hPrt1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human EIF3B fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8662
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.