Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3C Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB10511A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10511A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10511A532 Supplier Novus Biologicals Supplier No. NB100511AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EIF3C Polyclonal specifically detects EIF3C in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen EIF3C
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cell migration-inducing protein 17, eIF3 p110, eIF3c, EIF3CL, eIF3-p110, EIF3S8FLJ55450, Eukaryotic translation initiation factor 3 subunit 8, eukaryotic translation initiation factor 3 subunit C, eukaryotic translation initiation factor 3, subunit 8 (110kD), eukaryotic translation initiation factor 3, subunit 8, 110kDa, eukaryotic translation initiation factor 3, subunit C, FLJ53378, FLJ54400, FLJ54404, FLJ55750, FLJ78287, MGC189737, MGC189744
Gene Symbols EIF3C
Host Species Rabbit
Immunogen A synthetic peptide derived from a sequence between residues 850 and 918 (the C-terminus) of human eukaryotic translation initiation factor 3, subunit 8, 110kDa using the numbering given in Swiss-Prot entry Q99613 (GeneID 8663).
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8663
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.