Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF3e Antibody, Novus Biologicals™
SDP

Catalog No. NB11085528 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11085528 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11085528 Supplier Novus Biologicals Supplier No. NB11085528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF3e Polyclonal specifically detects eIF3e in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Knockdown Validated.

Specifications

Antigen eIF3e
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.8 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:90000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P60229
Gene Alias eIF-3 p48, eIF3e, eIF3-p48, EIF3S6EIF3-P48, Eukaryotic translation initiation factor 3 subunit 6, eukaryotic translation initiation factor 3 subunit E, eukaryotic translation initiation factor 3, subunit 6 (48kD), eukaryotic translation initiation factor 3, subunit 6 48kDa, eukaryotic translation initiation factor 3, subunit E, INT6eIF3-p46, mammary tumor-associated protein INT6, murine mammary tumor integration site 6 (oncogene homolog), Viral integration site protein INT-6 homolog
Gene Symbols EIF3E
Host Species Rabbit
Immunogen This affinity purified eIF3e Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminus of mouse eIF3e. (Uniprot: P60229)
Molecular Weight of Antigen 52.22 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3646
Test Specificity This is specific for mouse eIF3S6/Int6 protein. A BLAST analysis was used to suggest cross-reactivity with most eIF3Se/Int6 isoforms from mouse, human, rat, dog, bovine, and monkey based on 100% identity with the immunizing sequence.
Target Species Human, Mouse, Monkey, Bovine, Canine
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.