Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3G Rabbit anti-Human, Alexa Fluor 532, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB098768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB098768 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB098768 Supplier Novus Biologicals Supplier No. NBP299199AF532

Rabbit Polyclonal Antibody

EIF3G Polyclonal specifically detects EIF3G in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen EIF3G
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias eIF3 p42, eIF3 p44, eIF-3-delta, eIF3-delta, eIF3g, EIF3-P42, eIF3-p44, EIF3S4eIF-3 RNA-binding subunit, Eukaryotic translation initiation factor 3 RNA-binding subunit, Eukaryotic translation initiation factor 3 subunit 4, eukaryotic translation initiation factor 3 subunit G, eukaryotic translation initiation factor 3 subunit p42, eukaryotic translation initiation factor 3, subunit 4 (delta, 44kD), eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa, eukaryotic translation initiation factor 3, subunit G
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human EIF3G.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8666
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.