Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3J Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB097350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB097350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB097350 Supplier Novus Biologicals Supplier No. NBP297521IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EIF3J Polyclonal specifically detects EIF3J in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen EIF3J
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias eIF3 p35, eIF3-alpha, eIF3j, eIF3-p35, EIF3S1eIF-3-alpha, Eukaryotic translation initiation factor 3 subunit 1, eukaryotic translation initiation factor 3 subunit J, eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD), eukaryotic translation initiation factor 3, subunit 1 alpha, 35kDa, eukaryotic translation initiation factor 3, subunit J
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human EIF3J fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8669
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.