Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3M Antibody, Novus Biologicals™
SDP

Catalog No. NB1002420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002420 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002420 Supplier Novus Biologicals Supplier No. NB1002420
Add to Cart
Add to Cart

Goat Polyclonal Antibody

EIF3M Polyclonal specifically detects EIF3M in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen EIF3M
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006351.2
Gene Alias B5 receptor, dendritic cell protein, eIF3m, eukaryotic translation initiation factor 3 subunit M, eukaryotic translation initiation factor 3, subunit M, Fetal lung protein B5, FLJ29030, GA17, hfl-B5, HFLB5, PCI domain containing 1 (herpesvirus entry mediator), PCI domain-containing protein 1, PCID1B5
Gene Symbols EIF3M
Host Species Goat
Immunogen Peptide with sequence C-NAWKQNLNKVKN corresponding to C-Terminus according to NP_006351.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10480
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.