Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF3X Antibody, Novus Biologicals™
SDP

Catalog No. NB10093306 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093306 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093306 Supplier Novus Biologicals Supplier No. NB10093306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

eIF3X Polyclonal specifically detects eIF3X in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, RNA immunoprecipitation, Western Blot.

Specifications

Antigen eIF3X
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, RNA Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-5 mcg/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O75153
Gene Alias CLU1, DKFZp686L05242, DKFZp686M09233, FLJ23672, hypothetical protein LOC23277, KIAA0664
Gene Symbols CLUH
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1259 and 1309 of human putative eukaryotic translation initiation factor 3 subunit using the numbering given in entry NP_056044.3 (GeneID 23277).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23277
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey and Gorilla. Mouse reactivity reported in scientific literature (PMID: 25349259).
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.