Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4A1 Antibody (7C3), Novus Biologicals™
SDP

Catalog No. NB165559 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165559 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165559 Supplier Novus Biologicals Supplier No. NBP315220
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

eIF4A1 Monoclonal antibody specifically detects eIF4A1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen eIF4A1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 7C3
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS (pH 7.4), 150mM NaCl, and 50% glycerol.
Gene Alias ATP-dependent RNA helicase eIF4A-1, DDX2Aeukaryotic initiation factor 4A-I, EC 3.6.1, EC 3.6.4.13, EIF4A, EIF-4A, eIF-4A-I, eIF4A-I, eukaryotic initiation factor 4AI, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A, isoform 1, eukaryotic translation initiation factor 4A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human eIF4A1. UniProt ID (P60842).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, mTOR Pathway, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1973
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.