Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF4A1 Rabbit anti-Human, Mouse, Rat, Clone: 7D5K2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315406100UL
Description
eIF4A1 Monoclonal antibody specifically detects eIF4A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| eIF4A1 | |
| Monoclonal | |
| Unconjugated | |
| ATP-dependent RNA helicase eIF4A-1, DDX2Aeukaryotic initiation factor 4A-I, EC 3.6.1, EC 3.6.4.13, EIF4A, EIF-4A, eIF-4A-I, eIF4A-I, eukaryotic initiation factor 4AI, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A, isoform 1, eukaryotic translation initiation factor 4A1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human eIF4A1 (P60842). | |
| 100 μg | |
| Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
| 1973 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| 7D5K2 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction