Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF4A1 Rabbit anti-Human, Mouse, Rat, Clone: 7D5K2, Novus Biologicals™

Rabbit Monoclonal Antibody
$479.00
Specifications
Antigen | eIF4A1 |
---|---|
Clone | 7D5K2 |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
eIF4A1 Monoclonal antibody specifically detects eIF4A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
eIF4A1 | |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1973 | |
IgG | |
Affinity purified |
7D5K2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ATP-dependent RNA helicase eIF4A-1, DDX2Aeukaryotic initiation factor 4A-I, EC 3.6.1, EC 3.6.4.13, EIF4A, EIF-4A, eIF-4A-I, eIF4A-I, eukaryotic initiation factor 4AI, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A, isoform 1, eukaryotic translation initiation factor 4A1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human eIF4A1 (P60842). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title