Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4A2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224529SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224529SS 0.025 mg
NBP224529 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224529SS Supplier Novus Biologicals Supplier No. NBP224529SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF4A2 Polyclonal specifically detects eIF4A2 in Human, Mouse, Canine, Chicken, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen eIF4A2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.025-0.05 ug/ml, Simple Western 1:200, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q14240
Gene Alias ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2
Gene Symbols EIF4A2
Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human eIF4A2 was used as the immunogen for the antibody.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1974
Test Specificity The immunogen for this antibody has 100% homology to eIF4A1 and 89% homology to eIF4A3. Specificity to eIF4A2 has not been determined.
Target Species Human, Mouse, Canine, Chicken, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.