Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF4A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
| Antigen | eIF4A2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.025-0.05 ug/ml, Simple Western 1:200, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224529SS
![]() |
Novus Biologicals
NBP224529SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224529
![]() |
Novus Biologicals
NBP224529 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
eIF4A2 Polyclonal specifically detects eIF4A2 in Human, Mouse, Canine, Chicken, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| eIF4A2 | |
| Western Blot 0.025-0.05 ug/ml, Simple Western 1:200, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cancer, mTOR Pathway | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2 | |
| EIF4A2 | |
| IgG | |
| Affinity Purified | |
| The immunogen for this antibody has 100% homology to eIF4A1 and 89% homology to eIF4A3. Specificity to eIF4A2 has not been determined. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Canine, Chicken, Primate | |
| Q14240 | |
| 1974 | |
| A portion of amino acids 50-100 of human eIF4A2 was used as the immunogen for the antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title