Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4A2 Antibody (PCRP-EIF4A2-2B5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB383850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383850 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383850 Supplier Novus Biologicals Supplier No. NBP308883V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eIF4A2 Monoclonal antibody specifically detects eIF4A2 in Human samples. It is validated for ELISA, Protein Array

Specifications

Antigen eIF4A2
Applications ELISA, Protein Array
Classification Monoclonal
Clone PCRP-EIF4A2-2B5
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2
Host Species Mouse
Immunogen Recombinant full-length human eIF4A2 protein (Uniprot: Q14240)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1974
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.