Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4B Antibody, Novus Biologicals™
SDP

Catalog No. NBP224475 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224475 0.1 mg
NBP224475SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224475 Supplier Novus Biologicals Supplier No. NBP224475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF4B Polyclonal specifically detects eIF4B in Human, Mouse, Rat, Bovine, Chicken, Primate samples. It is validated for Western Blot.

Specifications

Antigen eIF4B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.25-0.50 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P23588
Gene Alias EIF-4B, eukaryotic translation initiation factor 4B, PRO1843
Gene Symbols EIF4B
Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human eIF4B was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, mTOR Pathway, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1975
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey and chimpanzee, 83% homologous in dog and 72% homologous in rat and mouse.
Target Species Human, Mouse, Rat, Bovine, Chicken, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.