Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4B Antibody, Novus Biologicals™
SDP

Catalog No. NB10093308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093308 Supplier Novus Biologicals Supplier No. NB10093308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

eIF4B Polyclonal specifically detects eIF4B in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen eIF4B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P23588
Gene Alias EIF-4B, eukaryotic translation initiation factor 4B, PRO1843
Gene Symbols EIF4B
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 561 and 611 of human eukaryotic translation initiation factor 4B using the numbering given in entry NP_001408.2 (GeneID 1975).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, mTOR Pathway, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1975
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Mouse, Dog, Horse, Panda, Orangutan, Rhesus Monkey, Gorilla, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Small-eared galago, Thirteen-lined ground squirrel and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.