Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4E Antibody (PCRP-EIF4E-1D3), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB383837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383837 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383837 Supplier Novus Biologicals Supplier No. NBP308884V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eIF4E Monoclonal antibody specifically detects eIF4E in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen eIF4E
Applications Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-EIF4E-1D3
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CBP, eIF4E, eIF-4E, EIF4E1, EIF4EL1MGC111573, EIF4F, eIF-4F 25 kDa subunit, eukaryotic translation initiation factor 4E, eukaryotic translation initiation factor 4E-like 1, mRNA cap-binding protein
Host Species Mouse
Immunogen Recombinant full-length human eIF4E protein (Uniprot: P06730)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1977
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.