Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4E2 Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene
SDP

Catalog No. p-200010982
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1569 30 μL
50-167-3462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1569 Supplier OriGene Supplier No. TA502402S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EIF4E2 Monoclonal antibody specifically detects EIF4E2 in Human samples. It is validated for Western Blot

Specifications

Antigen EIF4E2
Applications Western Blot
Classification Monoclonal
Clone OTI1F5
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HAGHL
Gene Accession No. Q6PII5
Gene Alias 2700069E09Rik; 4EHP; 4E-LP; AI036339; AV129531; D0H0S6743E; EGK_04904; eIF4E type 2; eIF-4E type 2; Eif4e2; eIF4E-2; EIF4EL3; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; eukar; eukaryotic translation initiation factor 4E family member 2; eukaryotic translation initiation factor 4E homologous protein; eukaryotic translation initiation factor 4E like 3; eukaryotic translation initiation factor 4E member 2; eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E-like 3; h4EHP; IF4e; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 2; mRNA cap-binding protein type 3
Host Species Mouse
Immunogen Full length human recombinant protein of human HAGHL produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84264
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.