Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4EBP2 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP11927005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP11927005 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP11927005 Supplier Novus Biologicals Supplier No. NBP1192700.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF4EBP2 Polyclonal specifically detects eIF4EBP2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen eIF4EBP2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500
Formulation No buffer with Sodium Azide
Gene Alias 4EBP2, 4E-BP2, eIF4E-binding protein 2, eukaryotic translation initiation factor 4E binding protein 2, eukaryotic translation initiation factor 4E-binding protein 2, PHASII, phosphorylated, heat and acid stable regulated by insulin protein II
Gene Symbols EIF4EBP2
Host Species Rabbit
Immunogen Synthetic peptide derived from the sequence of human PHAS-II, conjugated to KLH; sequence identical to mouse
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 1979
Test Specificity eIF4E-BP2
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.