Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4G1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB100268H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100268H 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100268H Supplier Novus Biologicals Supplier No. NB100268H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF4G1 Polyclonal specifically detects eIF4G1 in Human, Pig samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen eIF4G1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias DKFZp686A1451, EIF4F, eIF-4G 1, EIF-4G1, EIF4-gamma, eIF-4-gamma 1, EIF4GI, EIF4Gp220, eucaryotic translation initiation factor 4G, eukaryotic translation initiation factor 4 gamma 1, eukaryotic translation initiation factor 4 gamma, 1, P220
Gene Symbols EIF4G1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 575 and 625 of human Eukaryotic Initiation Factor 4 Gamma 1 (NP_886553.2).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1981
Target Species Human, Pig
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.