Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4G1, p Ser1231 Antibody, Novus Biologicals™
SDP

Catalog No. NBP104965 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104965 0.1 mL
NB018275 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP104965 Supplier Novus Biologicals Supplier No. NBP104965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

eIF4G1 (p Ser1231) Polyclonal antibody specifically detects eIF4G1 (p Ser1231) in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen eIF4G1 (p Ser1231)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Formulation PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide
Gene Accession No. Q04637
Gene Alias DKFZp686A1451, EIF4F, eIF-4G 1, EIF-4G1, EIF4-gamma, eIF-4-gamma 1, EIF4GI, EIF4Gp220, eucaryotic translation initiation factor 4G, eukaryotic translation initiation factor 4 gamma 1, eukaryotic translation initiation factor 4 gamma, 1, P220
Gene Symbols EIF4G1
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human eIF4G1 [p Ser1231] around the phosphorylation site of serine 1231 (P-V-Sp-P-L).
Molecular Weight of Antigen 220 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1981
Test Specificity Detects endogenous levels of eIF4G1 only when phosphorylated at serine 1231.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.