Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF5A Antibody (4D1), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB160526 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160526 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160526 Supplier Novus Biologicals Supplier No. NBP259403JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

eIF5A Monoclonal antibody specifically detects eIF5A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen eIF5A
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 4D1
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias eIF-4D, eIF-5A, eIF-5A1, eIF-5A-1, EIF5A1, eIF5AI, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 1, eukaryotic translation initiation factor 5A, eukaryotic translation initiation factor 5A-1, MGC104255, MGC99547, Rev-binding factor
Host Species Mouse
Immunogen Recombinant human eIF5A (1-154aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 1984
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.