Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF5A2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB315709 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315709 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315709 Supplier Novus Biologicals Supplier No. NBP296987JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF5A2 Polyclonal antibody specifically detects eIF5A2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen eIF5A2
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias eIF-5A-2, EIF-5A2, eIF5AII, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 2, eukaryotic translation initiation factor 5A2, eukaryotic translation initiation factor 5A-2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human eIF5A2 (Uniprot#: Q9GZV4; Met1-Lys153)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 56648
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.