Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF5A2 Antibody (OTI6G7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB272056APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272056APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272056APC Supplier Novus Biologicals Supplier No. NBP272056APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eIF5A2 Monoclonal specifically detects eIF5A2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen eIF5A2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI6G7
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias eIF-5A-2, EIF-5A2, eIF5AII, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 2, eukaryotic translation initiation factor 5A2, eukaryotic translation initiation factor 5A-2
Gene Symbols EIF5A2
Host Species Mouse
Immunogen Full length recombinant protein of human eIF5A2 (NP_065123) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 56648.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.