Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIG121 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341751 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341751 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341751 Supplier Novus Biologicals Supplier No. NBP281949MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EIG121 Polyclonal antibody specifically detects EIG121 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen EIG121
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias estrogen-induced gene 121 protein, hypothetical protein LOC57535, KIAA1324, MGC150624
Host Species Rabbit
Immunogen EIG121 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human EIG121. The immunogen is located within the last 50 amino acids of EIG121. Amino Acid Squence: IKSFTSKRTPDGFDSVPL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57535
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.