Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Bar Magnets
SCI_ED

Catalog No. p-3649333 Shop All Eisco Products
Change view
Click to view available options
Color:
Red and Blue
Znc-plated steel
Shape:
Bar
Rectangular
Weight (English):
0.95 lb.
3.5 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Shape Weight (English)
S95965 Red and Blue Bar 3.5 lb.
S95966 Znc-plated steel Rectangular 0.95 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S95965 Supplier Eisco™ Supplier No. PH0786D
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Perfect for classroom experiments.

  • Red and blue magnets with stamped polarity measure 3.9 x 0.79 x 0.23 in. (10 x 2 x 0.6cm) and are boxed
  • Rectangular square-faced chrome steel magnets measure 5.9 x 0.4 x 0.4 in. (15 x 1 x 1cm)

Order Info

Magnets sold in packs of two.

Specifications

Material Chrome Steel
Shape Bar
For Use With (Application) Perfect for classroom experiments
Grade 9 to 12
Color Red and Blue
Product Type Bar Magnet
Includes Keeper
Weight (Metric) 1.59 kg
Width (English) 5.51 in.
Weight (English) 3.5 lb.
Width (Metric) 14 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.