Learn More
Description
Includes
Activity guide
- Discover how the use of pulleys increases mechanical advantage
- Durable wooden construction is ideal for hands-on classroom use
Specifications
Specifications
| Product Type | Block and Tackle Mode |
| Material | Wood |
| For Use With (Application) | Classroom use, lifts large loads with little effort |
| Includes | Activity guide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
