Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Tuning Forks Kit
SCI_ED

Catalog No. s02108 Shop All Eisco Products
Click to view available options
Class Size:
1 to 2 Lab Groups

Includes

  • Two wooden resonator boxes
  • Two tuning forks
  • Sliding weight
  • Hammer
  • Instructions

Requires

NeuLog™ Sound Logger Sensor (S98243-22ND)

The Tuning Forks Kit demonstrates relation between sound and wave propagation.

Students produce sound waves generated by tuning forks and measure them using a separate sound sensor.

Kit enables students to study:

  • Sound
  • Sound waves
  • Sound beats

Specifications

Grade 6 to Undergraduate
For Use With (Application) Demonstrates relation between sound and wave propagation
Includes Two wooden resonator boxes, two tuning forks, sliding weight, hammer, instructions
Class Size 1 to 2 Lab Groups
Product Type Tuning Forks Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.